accessibility menu, dialog, popup

UserName

CAS RN 61040-78-6

IUPAC Name: (2,4,6-trimethoxyphenyl)methanol
Synonyms: (2,4,6-trimethoxyphenyl)methanol
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: CFXXBVNHYJQNKS-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(CO)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results