missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzonitrile, 98%

Catalog No. p-7027072
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2354006 5 g
AAB2354014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2354006 Supplier Thermo Scientific Chemicals Supplier No. B2354006

CAS: 2571-54-2 | C10H11NO3 | 193.202 g/mol

2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 2571-54-2
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD00001787
InChI Key GBRHJUMDNWLSCT-UHFFFAOYSA-N
PubChem CID 75731
IUPAC Name 2,4,6-trimethoxybenzonitrile
SMILES COC1=CC(=C(C(=C1)OC)C#N)OC

Specifications

Melting Point 140°C to 143°C
Odor Characteristic
Quantity 5 g
Synonym benzonitrile, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzonitrile, 2,4,6-trimethoxybenzenecarbonitrile, acmc-20aoeu, 2,4,6-trimethoxy benzonitrile, benzonitrile,2,4,6-trimethoxy, 2,4,6-trimethoxybenzonitrile
Solubility Information Insoluble in water.
Formula Weight 193.2
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethoxybenzonitrile
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 219-917-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only