accessibility menu, dialog, popup

UserName

CAS RN 2571-54-2

IUPAC Name: 2,4,6-trimethoxybenzonitrile
Synonyms: 2,4,6-trimethoxybenzonitrile
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: GBRHJUMDNWLSCT-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C#N)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results