missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethoxybenzoic Acid 98.0+%, TCI America™
$652.64 - $652.64
Chemical Identifiers
| CAS | 570-02-5 |
|---|---|
| Molecular Formula | C10H12O5 |
| Molecular Weight (g/mol) | 212.20 |
| MDL Number | MFCD00016497 |
| InChI Key | JATAKEDDMQNPOQ-UHFFFAOYSA-N |
| Synonym | benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid |
| PubChem CID | 68441 |
| IUPAC Name | 2,4,6-trimethoxybenzoic acid |
| SMILES | COC1=CC(OC)=C(C(O)=O)C(OC)=C1 |
Chemical Identifiers
| 570-02-5 | |
| 212.20 | |
| JATAKEDDMQNPOQ-UHFFFAOYSA-N | |
| 68441 | |
| COC1=CC(OC)=C(C(O)=O)C(OC)=C1 |
| C10H12O5 | |
| MFCD00016497 | |
| benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid | |
| 2,4,6-trimethoxybenzoic acid |
Specifications
| 570-02-5 | |
| White-Yellow | |
| MFCD00016497 | |
| benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid | |
| COC1=CC(OC)=C(C(O)=O)C(OC)=C1 | |
| 212.20 | |
| 212.20 | |
| Crystalline Powder |
| 146°C | |
| C10H12O5 | |
| 5 g | |
| JATAKEDDMQNPOQ-UHFFFAOYSA-N | |
| 2,4,6-trimethoxybenzoic acid | |
| 68441 | |
| ≥98.0% (GC,T) | |
| 2,4,6-Trimethoxybenzoic Acid |
Safety and Handling
TSCA : Yes