accessibility menu, dialog, popup

UserName

H3COOCH3OHOOH3C

CAS RN 570-02-5

IUPAC Name: 2,4,6-trimethoxybenzoic acid
Synonyms: 2,4,6-trimethoxybenzoic acid
Molecular Weight (g/mol): 212.2
Molecular Formula: C10H12O5
InChi Key: JATAKEDDMQNPOQ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C(O)=O)C(OC)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)