missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. T24085G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24085G Supplier TCI America Supplier No. T24085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 570-02-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.20
MDL Number MFCD00016497
InChI Key JATAKEDDMQNPOQ-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid
PubChem CID 68441
IUPAC Name 2,4,6-trimethoxybenzoic acid
SMILES COC1=CC(OC)=C(C(O)=O)C(OC)=C1

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 5 g
Formula Weight 212.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethoxybenzoic Acid