missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzoic acid, 98%

Catalog No. AAL0499706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0499706 5 g
AAL0499714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0499706 Supplier Thermo Scientific Chemicals Supplier No. L0499706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 570-02-5 | C10H12O5 | 212.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 570-02-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.20
MDL Number MFCD00016497
InChI Key JATAKEDDMQNPOQ-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid
PubChem CID 68441
IUPAC Name 2,4,6-trimethoxybenzoic acid
SMILES COC1=CC(OC)=C(C(O)=O)C(OC)=C1

Specifications

Melting Point ∽158°C (decomposition)
Quantity 5 g
Beilstein 1972294
Formula Weight 212.2
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-325-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only