missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triisopropylbenzenesulfonyl hydrazide
Description
As a reagent for generation of diimide
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 110°C to 112°C (decomposition) |
| Quantity | 0.5 g |
| Beilstein | 2145001 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 298.58 |
| Physical Form | Crystalline |
| Chemical Name or Material | 2, 4, 6-Triisopropylbenzenesulfonyl hydrazide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 254-282-7 |
| TSCA | No |
| storageNote1 | Moisture Sensitive |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only