missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonamide, 98%

Catalog No. aal0844514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0844514 25 g
AAL0844506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0844514 Supplier Thermo Scientific Chemicals Supplier No. L0844514
Only null left

CAS: 105536-22-9 | C15H25NO2S | 283.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105536-22-9
Molecular Formula C15H25NO2S
Molecular Weight (g/mol) 283.43
MDL Number MFCD00051975
InChI Key PRMNQLMPSVOZIX-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonamide, 2,4,6-tri propan-2-yl benzenesulfonamide, 2,4,6-tris methylethyl benzenesulfonamide, cbmicro_034663, acmc-1bt32, 2,4,6-triisopropylbenzenesulfonamine, benzenesulfonamide,2,4,6-tris 1-methylethyl
PubChem CID 736244
IUPAC Name 2,4,6-tri(propan-2-yl)benzenesulfonamide
SMILES CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(N)(=O)=O

Specifications

Melting Point 113°C to 116°C
Quantity 25 g
Beilstein 2658523
Formula Weight 283.44
Percent Purity 98%
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only