CAS RN 105536-22-9
IUPAC Name:
2,4,6-tris(propan-2-yl)benzene-1-sulfonamide
Synonyms:
2,4,6-triisopropylbenzenesulfonamide
Molecular Weight (g/mol):
283.43
Molecular Formula:
C15H25NO2S
InChi Key:
PRMNQLMPSVOZIX-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(N)(=O)=O
1
–
1
of
1
results
1
Promotions Available
2,4,6-Triisopropylbenzenesulfonamide, 98%
2,4,6-Triisopropylbenzenesulfonamide, 98%
CAS: 105536-22-9 Molecular Formula: C15H25NO2S Molecular Weight (g/mol): 283.43 MDL Number: MFCD00051975 InChI Key: PRMNQLMPSVOZIX-UHFFFAOYSA-N Synonym: 2,4,6-triisopropylbenzenesulfonamide,2,4,6-tri propan-2-yl benzenesulfonamide,2,4,6-tris methylethyl benzenesulfonamide,cbmicro_034663,acmc-1bt32,2,4,6-triisopropylbenzenesulfonamine,benzenesulfonamide,2,4,6-tris 1-methylethyl PubChem CID: 736244 IUPAC Name: 2,4,6-tri(propan-2-yl)benzenesulfonamide SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(N)(=O)=O