missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trihydroxyacetophenone, 98%, Thermo Scientific™

Chemical Identifiers

CAS 480-66-0
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00149091,MFCD00002287
InChI Key XLEYFDVVXLMULC-UHFFFAOYSA-N
Synonym 1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone
PubChem CID 68073
ChEBI CHEBI:64344
IUPAC Name 1-(2,4,6-trihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(O)C=C1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC139790050
SDS View Documents
Thermo Scientific Chemicals
139790050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC139790250
SDS View Documents
Thermo Scientific Chemicals
139790250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC139791000
SDS View Documents
Thermo Scientific Chemicals
139791000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

480-66-0
168.15
XLEYFDVVXLMULC-UHFFFAOYSA-N
68073
1-(2,4,6-trihydroxyphenyl)ethanone
C8H8O4
MFCD00149091,MFCD00002287
1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone
CHEBI:64344
CC(=O)C1=C(O)C=C(O)C=C1O

Specifications

480-66-0
97.5% min. (HPLC)
C8H8O4
MFCD00149091,MFCD00002287
08,II,442
XLEYFDVVXLMULC-UHFFFAOYSA-N
1-(2,4,6-trihydroxyphenyl)ethanone
68073
168.15
2', 4', 6'-Trihydroxyacetophenone
Authentic
Glass bottle
(HO)3C6H2COCH3
5g
1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone
CC(=O)C1=C(O)C=C(O)C=C1O
168.15
CHEBI:64344
98%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 207-556-5

SDS
Documents

Documents

RUO – Research Use Only