accessibility menu, dialog, popup

UserName

CAS RN 480-66-0

H3COOHOHHO

CAS RN 480-66-0

IUPAC Name: 1-(2,4,6-trihydroxyphenyl)ethan-1-one
Synonyms: phloroacetophenone|2,4,6-trihydroxyacetophenone
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: XLEYFDVVXLMULC-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C=C(O)C=C1O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)