missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4',6'-Trihydroxyacetophenone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 139791000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2', 4', 6'-Trihydroxyacetophenone | |
| Authentic | |
| Plastic Bottle | |
| (HO)3C6H2COCH3 | |
| 100g | |
| 1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone | |
| CC(=O)C1=C(O)C=C(O)C=C1O | |
| 168.15 | |
| CHEBI:64344 | |
| 98% |
| 480-66-0 | |
| 97.5% min. (HPLC) | |
| C8H8O4 | |
| MFCD00149091,MFCD00002287 | |
| 08,II,442 | |
| XLEYFDVVXLMULC-UHFFFAOYSA-N | |
| 1-(2,4,6-trihydroxyphenyl)ethanone | |
| 68073 | |
| 168.15 |
Chemical Identifiers
| 480-66-0 | |
| 168.15 | |
| XLEYFDVVXLMULC-UHFFFAOYSA-N | |
| 68073 | |
| 1-(2,4,6-trihydroxyphenyl)ethanone |
| C8H8O4 | |
| MFCD00149091,MFCD00002287 | |
| 1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone | |
| CHEBI:64344 | |
| CC(=O)C1=C(O)C=C(O)C=C1O |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 207-556-5
RUO â Research Use Only