missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorophenylboronic acid, 95%

Catalog No. AC433590010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433590010 1 g Glass bottle
AC433590050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433590010 Supplier Thermo Scientific Chemicals Supplier No. 433590010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 182482-25-3 | C6H4BF3O2 | 175.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 182482-25-3
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.90
MDL Number MFCD01863169
InChI Key IPEIGKHHSZFAEW-UHFFFAOYSA-N
Synonym 2,4,6-trifluorophenyl boronic acid, 2,4,6-trifluorobenzeneboronic acid, 2,4,6-trifluorophenylboronicacid, 2,4,6-trifluorophenyl boranediol, boronic acid, 2,4,6-trifluorophenyl, pubchem1788, acmc-209ejj, chembl91770, boronicacid, b-2,4,6-trifluorophenyl, boronic acid, b-2,4,6-trifluorophenyl
PubChem CID 2779329
IUPAC Name (2,4,6-trifluorophenyl)boronic acid
SMILES OB(O)C1=C(F)C=C(F)C=C1F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 239.5°C to 247.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 175.9
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trifluorophenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only