accessibility menu, dialog, popup

UserName

HOBOHFFF

CAS RN 182482-25-3

IUPAC Name: (2,4,6-trifluorophenyl)boronic acid
Synonyms: 2,4,6-trifluorophenylboronic acid
Molecular Weight (g/mol): 175.9
Molecular Formula: C6H4BF3O2
InChi Key: IPEIGKHHSZFAEW-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=C(F)C=C1F
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results