Learn More
2,4,6-Trifluorobenzoyl chloride, 97%
CAS: 79538-29-7 | C7H2ClF3O | 194.537 g/mol
Supplier: Thermo Scientific Chemicals B2032306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,4,6-Trifluorobenzoyl chloride | |
| 1.499 | |
| C7H2ClF3O | |
| MFCD00070810 | |
| UN1760 | |
| Moisture sensitive | |
| SIFIJQFBERMWMU-UHFFFAOYSA-N | |
| 2,4,6-trifluorobenzoyl chloride | |
| 144895 | |
| 97% |
| 79538-29-7 | |
| 167°C to 168°C | |
| 1.484 | |
| 5 g | |
| 1958563 | |
| 2,4,6-trifluorobenzoylchloride, 2,4,6-trifluoro-benzoyl chloride, acmc-20aof3, timtec-bb sbb005827, 2,4,6-tri-fluorobenzoyl chloride, 2,4,6-trifluorobenzoyl chloride, benzoyl chloride,2,4,6-trifluoro, benzoyl chloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzoyl chloride 1g, benzoyl chloride, 2,4,6-trifluoro-9ci | |
| C1=C(C=C(C(=C1F)C(=O)Cl)F)F | |
| 194.537 | |
| 194.54 |
Chemical Identifiers
| 79538-29-7 | |
| 194.537 | |
| SIFIJQFBERMWMU-UHFFFAOYSA-N | |
| 144895 | |
| C1=C(C=C(C(=C1F)C(=O)Cl)F)F |
| C7H2ClF3O | |
| MFCD00070810 | |
| 2,4,6-trifluorobenzoylchloride, 2,4,6-trifluoro-benzoyl chloride, acmc-20aof3, timtec-bb sbb005827, 2,4,6-tri-fluorobenzoyl chloride, 2,4,6-trifluorobenzoyl chloride, benzoyl chloride,2,4,6-trifluoro, benzoyl chloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzoyl chloride 1g, benzoyl chloride, 2,4,6-trifluoro-9ci | |
| 2,4,6-trifluorobenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only