missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichlorophenol 97.0+%, TCI America™
$42.87 - $193.86
Chemical Identifiers
| CAS | 88-06-2 |
|---|---|
| Molecular Formula | C6H3Cl3O |
| Molecular Weight (g/mol) | 197.439 |
| MDL Number | MFCD00002172 |
| InChI Key | LINPIYWFGCPVIE-UHFFFAOYSA-N |
| Synonym | dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz |
| PubChem CID | 6914 |
| ChEBI | CHEBI:28755 |
| IUPAC Name | 2,4,6-trichlorophenol |
| SMILES | C1=C(C=C(C(=C1Cl)O)Cl)Cl |
Chemical Identifiers
| 88-06-2 | |
| 197.439 | |
| LINPIYWFGCPVIE-UHFFFAOYSA-N | |
| 6914 | |
| 2,4,6-trichlorophenol |
| C6H3Cl3O | |
| MFCD00002172 | |
| dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz | |
| CHEBI:28755 | |
| C1=C(C=C(C(=C1Cl)O)Cl)Cl |
Specifications
| 88-06-2 | |
| White-Yellow | |
| C6H3Cl3O | |
| 25 g | |
| dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz | |
| C1=C(C=C(C(=C1Cl)O)Cl)Cl | |
| 197.439 | |
| CHEBI:28755 | |
| ≥97.0% (GC) | |
| 2,4,6-Trichlorophenol |
| 67°C | |
| 120°C | |
| MFCD00002172 | |
| 2020 | |
| LINPIYWFGCPVIE-UHFFFAOYSA-N | |
| 2,4,6-trichlorophenol | |
| 6914 | |
| 197.44 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0931
RTECSNumber : SN1575000
TSCA : Yes