accessibility menu, dialog, popup

UserName

CAS RN 88-06-2

IUPAC Name: 2,4,6-trichlorophenol
Synonyms: 2,4,6-trichlorophenol phenachlor OMAL 2,4,6-trichlorfenol 2,4,6-trichlorophenol, zn salt phenaclor ccris 605 rcra waste number U231 dowicide 2S zn salt 2,4,6-trichlorophenol
Molecular Weight (g/mol): 197.44
Molecular Formula: C6H3Cl3O
InChi Key: LINPIYWFGCPVIE-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(Cl)C=C1Cl
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)