missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorophenol 97.0+%, TCI America™

Catalog No. T039025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T039025G 25 g
T0390500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T039025G Supplier TCI America Supplier No. T039025G

Chemical Identifiers

CAS 88-06-2
Molecular Formula C6H3Cl3O
Molecular Weight (g/mol) 197.439
MDL Number MFCD00002172
InChI Key LINPIYWFGCPVIE-UHFFFAOYSA-N
Synonym dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz
PubChem CID 6914
ChEBI CHEBI:28755
IUPAC Name 2,4,6-trichlorophenol
SMILES C1=C(C=C(C(=C1Cl)O)Cl)Cl

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 120°C
Quantity 25 g
UN Number 2020
Formula Weight 197.44
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trichlorophenol
EINECSNumber (3)-0931
RTECSNumber SN1575000
TSCA Yes