missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7136816
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T151810G 10 g
T151825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T151810G Supplier TCI America Supplier No. T151810G
Only null left

Chemical Identifiers

CAS 50-43-1
Molecular Formula C7H3Cl3O2
Molecular Weight (g/mol) 225.449
MDL Number MFCD00060699
InChI Key RAFFVQBMVYYTQS-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trichloro, unii-0bwg1hx6ck, 2,4,6-trichlorobenzoicacid, 2,4,6-trichloro-benzoic acid, 0bwg1hx6ck, 2,4,6-trichlorbenzoes, pubchem18496, acmc-209kme, 2,4,6-trichlorobbenzoicacid, ksc270e7f
PubChem CID 5764
IUPAC Name 2,4,6-trichlorobenzoic acid
SMILES C1=C(C=C(C(=C1Cl)C(=O)O)Cl)Cl

Specifications

Melting Point 165°C
Quantity 10 g
Formula Weight 225.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trichlorobenzoic Acid
EINECSNumber (3)-0062
RTECSNumber DH8050000
TSCA No