accessibility menu, dialog, popup

UserName

CAS RN 50-43-1

IUPAC Name: 2,4,6-trichlorobenzoate
Synonyms: 2,4,6-trichlorobenzoate
Molecular Weight (g/mol): 224.44
Molecular Formula: C7H2Cl3O2
InChi Key: RAFFVQBMVYYTQS-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(Cl)C=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)