missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorobenzoic acid, 94%

Catalog No. AAL1419106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1419106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1419106 Supplier Thermo Scientific Chemicals Supplier No. L1419106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50-43-1 | C7H3Cl3O2 | 225.449 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-43-1
Molecular Formula C7H3Cl3O2
Molecular Weight (g/mol) 225.449
MDL Number MFCD00060699
InChI Key RAFFVQBMVYYTQS-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trichloro, unii-0bwg1hx6ck, 2,4,6-trichlorobenzoicacid, 2,4,6-trichloro-benzoic acid, 0bwg1hx6ck, 2,4,6-trichlorbenzoes, pubchem18496, acmc-209kme, 2,4,6-trichlorobbenzoicacid, ksc270e7f
PubChem CID 5764
IUPAC Name 2,4,6-trichlorobenzoic acid
SMILES C1=C(C=C(C(=C1Cl)C(=O)O)Cl)Cl

Specifications

Melting Point 162°C to 164°C
Quantity 5 g
Beilstein 1874127
Formula Weight 225.46
Percent Purity 94%
Chemical Name or Material 2,4,6-Trichlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DH8050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only