missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tribromoanisole 98.0+%, TCI America™
Supplier: TCI America T230125G
Specifications
| 2,4,6-Tribromoanisole | |
| 88°C | |
| 299°C | |
| MFCD00192510 | |
| 2,4,6-tribromoanisole, anisole, 2,4,6-tribromo, benzene, 1,3,5-tribromo-2-methoxy, unii-do7m3m4lx5, 1,3,5-tribromo-2-methoxy-benzene, methyl 2,4,6-tribromophenyl ether, do7m3m4lx5, 2,6-tribromoanisole, anisole,4,6-tribromo, acmc-209mm5 | |
| COC1=C(Br)C=C(Br)C=C1Br | |
| 344.83 | |
| 344.83 | |
| Crystalline Powder |
| 607-99-8 | |
| White-Yellow | |
| C7H5Br3O | |
| 25 g | |
| YXTRCOAFNXQTKL-UHFFFAOYSA-N | |
| 1,3,5-tribromo-2-methoxybenzene | |
| 11839 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 607-99-8 | |
| 344.83 | |
| YXTRCOAFNXQTKL-UHFFFAOYSA-N | |
| 11839 | |
| COC1=C(Br)C=C(Br)C=C1Br |
| C7H5Br3O | |
| MFCD00192510 | |
| 2,4,6-tribromoanisole, anisole, 2,4,6-tribromo, benzene, 1,3,5-tribromo-2-methoxy, unii-do7m3m4lx5, 1,3,5-tribromo-2-methoxy-benzene, methyl 2,4,6-tribromophenyl ether, do7m3m4lx5, 2,6-tribromoanisole, anisole,4,6-tribromo, acmc-209mm5 | |
| 1,3,5-tribromo-2-methoxybenzene |
Safety and Handling
TSCA : Yes