missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromoanisole 98.0+%, TCI America™

Catalog No. p-7137149
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T23015G 5 g
T230125G 25 g
2 options

Catalog No. T23015G

Supplier: TCI America T23015G

Only null left

Chemical Identifiers

CAS 607-99-8
Molecular Formula C7H5Br3O
Molecular Weight (g/mol) 344.83
MDL Number MFCD00192510
InChI Key YXTRCOAFNXQTKL-UHFFFAOYSA-N
Synonym 2,4,6-tribromoanisole, anisole, 2,4,6-tribromo, benzene, 1,3,5-tribromo-2-methoxy, unii-do7m3m4lx5, 1,3,5-tribromo-2-methoxy-benzene, methyl 2,4,6-tribromophenyl ether, do7m3m4lx5, 2,6-tribromoanisole, anisole,4,6-tribromo, acmc-209mm5
PubChem CID 11839
IUPAC Name 1,3,5-tribromo-2-methoxybenzene
SMILES COC1=C(Br)C=C(Br)C=C1Br

Specifications

Melting Point 88°C
Color White-Yellow
Boiling Point 299°C
Quantity 5 g
Formula Weight 344.83
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tribromoanisole