missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Trimethoxybenzaldehyde 98.0+%, TCI America™
$29.83 - $85.75
Chemical Identifiers
| CAS | 4460-86-0 |
|---|---|
| Molecular Formula | C10H12O4 |
| Molecular Weight (g/mol) | 196.20 |
| MDL Number | MFCD00003312 |
| InChI Key | IAJBQAYHSQIQRE-UHFFFAOYSA-N |
| Synonym | asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296 |
| PubChem CID | 20525 |
| IUPAC Name | 2,4,5-trimethoxybenzaldehyde |
| SMILES | COC1=CC(OC)=C(C=O)C=C1OC |
Chemical Identifiers
| 4460-86-0 | |
| 196.20 | |
| IAJBQAYHSQIQRE-UHFFFAOYSA-N | |
| 20525 | |
| COC1=CC(OC)=C(C=O)C=C1OC |
| C10H12O4 | |
| MFCD00003312 | |
| asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296 | |
| 2,4,5-trimethoxybenzaldehyde |
Specifications
| 4460-86-0 | |
| Yellow | |
| C10H12O4 | |
| 5 g | |
| IAJBQAYHSQIQRE-UHFFFAOYSA-N | |
| 2,4,5-trimethoxybenzaldehyde | |
| 20525 | |
| ≥98.0% (GC) | |
| 2,4,5-Trimethoxybenzaldehyde |
| 113°C | |
| 140°C | |
| MFCD00003312 | |
| asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296 | |
| COC1=CC(OC)=C(C=O)C=C1OC | |
| 196.20 | |
| 196.20 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CU8460000
TSCA : Yes