accessibility menu, dialog, popup

UserName

CAS RN 4460-86-0

IUPAC Name: 2,4,5-trimethoxybenzaldehyde
Synonyms: 2,4,5-trimethoxybenzaldehyde
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: IAJBQAYHSQIQRE-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C=O)C=C1OC
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)