missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethoxybenzaldehyde, 98%

Catalog No. p-206432
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157940250 25 g Glass bottle
AC157941000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157940250 Supplier Thermo Scientific Chemicals Supplier No. 157940250
Only null left

CAS: 4460-86-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4460-86-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00003312
InChI Key IAJBQAYHSQIQRE-UHFFFAOYSA-N
Synonym asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296
PubChem CID 20525
IUPAC Name 2,4,5-trimethoxybenzaldehyde
SMILES COC1=CC(OC)=C(C=O)C=C1OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 111.0°C to 115.0°C
Color White to Beige
Boiling Point 140°C (4.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3O)3C6H2CHO
Quantity 25 g
Beilstein 08, 389
Formula Weight 196.2
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2, 4, 5-Trimethoxybenzaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 224-713-3
RTECSNumber CU8460000
TSCA TSCA

RUO – Research Use Only