missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethoxybenzaldehyde, 98%

Catalog No. AAB2279214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2279214 25 g
AAB2279222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2279214 Supplier Thermo Scientific Chemicals Supplier No. B2279214

CAS: 4460-86-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4460-86-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00003312
InChI Key IAJBQAYHSQIQRE-UHFFFAOYSA-N
Synonym asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296
PubChem CID 20525
IUPAC Name 2,4,5-trimethoxybenzaldehyde
SMILES COC1=CC(OC)=C(C=O)C=C1OC

Specifications

Melting Point 111°C to 115°C
Boiling Point 140°C (4 mmHg)(sublimation)
Quantity 25 g
Beilstein 1951403
Sensitivity Air Sensitive
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 2,4,5-Trimethoxybenzaldehyde
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-713-3
RTECSNumber CU8460000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only