missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Trifluorobenzoic Acid 97.0+%, TCI America™
Supplier: TCI America T158625G
Specifications
| 2,4,5-Trifluorobenzoic Acid | |
| 98°C | |
| C7H2F3O2 | |
| 25 g | |
| AKAMNXFLKYKFOJ-UHFFFAOYSA-M | |
| 2,4,5-trifluorobenzoate | |
| 521170 | |
| ≥97.0% (GC,T) |
| 446-17-3 | |
| White | |
| MFCD00013306 | |
| 2,4,5-trifluoro-benzoic acid, benzoic acid, 2,4,5-trifluoro, pubchem1346, acmc-209jyu, 2,4,5-trifluorobenzoicacid, ksc235o3j, 2,4,5-trifluoro benzoic acid, rarechem al bo 0258, akamnxflkykfoj-uhfffaoysa, 2,4,5-trifluorobenzoic acid | |
| [O-]C(=O)C1=CC(F)=C(F)C=C1F | |
| 175.09 | |
| 176.09 | |
| Crystalline Powder |
Chemical Identifiers
| 446-17-3 | |
| 175.09 | |
| AKAMNXFLKYKFOJ-UHFFFAOYSA-M | |
| 521170 | |
| [O-]C(=O)C1=CC(F)=C(F)C=C1F |
| C7H2F3O2 | |
| MFCD00013306 | |
| 2,4,5-trifluoro-benzoic acid, benzoic acid, 2,4,5-trifluoro, pubchem1346, acmc-209jyu, 2,4,5-trifluorobenzoicacid, ksc235o3j, 2,4,5-trifluoro benzoic acid, rarechem al bo 0258, akamnxflkykfoj-uhfffaoysa, 2,4,5-trifluorobenzoic acid | |
| 2,4,5-trifluorobenzoate |
Safety and Handling
TSCA : No