missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzoic Acid 97.0+%, TCI America™

Catalog No. p-7136854
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15865G 5 g
T158625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15865G Supplier TCI America Supplier No. T15865G
Only null left

Chemical Identifiers

CAS 446-17-3
Molecular Formula C7H2F3O2
Molecular Weight (g/mol) 175.09
MDL Number MFCD00013306
InChI Key AKAMNXFLKYKFOJ-UHFFFAOYSA-M
Synonym 2,4,5-trifluoro-benzoic acid, benzoic acid, 2,4,5-trifluoro, pubchem1346, acmc-209jyu, 2,4,5-trifluorobenzoicacid, ksc235o3j, 2,4,5-trifluoro benzoic acid, rarechem al bo 0258, akamnxflkykfoj-uhfffaoysa, 2,4,5-trifluorobenzoic acid
PubChem CID 521170
IUPAC Name 2,4,5-trifluorobenzoate
SMILES [O-]C(=O)C1=CC(F)=C(F)C=C1F

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 176.09
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trifluorobenzoic Acid