missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Trifluoromethylphenyl)ethylamine 96.0+%, TCI America™

Catalog No. p-7137568
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30861G 1 g
T30865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30861G Supplier TCI America Supplier No. T30861G

Chemical Identifiers

CAS 52516-30-0
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD00040756
InChI Key BPVYCXMGJPKOTQ-UHFFFAOYSA-N
Synonym 2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
PubChem CID 104223
IUPAC Name 2-[3-(trifluoromethyl)phenyl]ethanamine
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCN

Specifications

Color Yellow
Quantity 1 g
UN Number 2735
Formula Weight 189.18
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(3-Trifluoromethylphenyl)ethylamine
RTECSNumber SI2525000
TSCA No