accessibility menu, dialog, popup

UserName

NH3FFF

CAS RN 52516-30-0

IUPAC Name: 2-[3-(trifluoromethyl)phenyl]ethan-1-aminium
Synonyms: 2-[3-(trifluoromethyl)phenyl]ethanaminium
Molecular Weight (g/mol): 190.19
Molecular Formula: C9H11F3N
InChi Key: BPVYCXMGJPKOTQ-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (7)
  • (3)
  • (1)
  • (3)
  • (3)