missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Trifluoromethyl)phenyl]ethylamine, 98%

Catalog No. AAH6408414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6408414 25 g
AAH6408403 1 g
AAH6408406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6408414 Supplier Thermo Scientific Chemicals Supplier No. H6408414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52516-30-0 | C9H10F3N | 189.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52516-30-0
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD00040756
InChI Key BPVYCXMGJPKOTQ-UHFFFAOYSA-N
Synonym 2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
PubChem CID 104223
IUPAC Name 2-[3-(trifluoromethyl)phenyl]ethanamine
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCN

Specifications

Color Colorless to Yellow
Density 1.18 g/mL
Boiling Point 113°C (22 mmHg)
Refractive Index 1.4645
Quantity 25 g
UN Number UN2735
Beilstein 4398117
Sensitivity Light sensitive
Formula Weight 189.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-[3-(Trifluoromethyl)phenyl]ethylamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 257-988-3
RTECSNumber SI2525000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only