missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Pentanedione 97.0+%, TCI America™
Supplier: TCI America P0051100ML
Specifications
| 2,3-Pentanedione | |
| -52°C | |
| 108°C | |
| MFCD00009313 | |
| 1224 | |
| TZMFJUDUGYTVRY-UHFFFAOYSA-N | |
| pentane-2,3-dione | |
| 11747 | |
| 100.12 | |
| Liquid |
| 600-14-6 | |
| Yellow | |
| C5H8O2 | |
| 100 mL | |
| 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone | |
| CCC(=O)C(C)=O | |
| 100.12 | |
| CHEBI:52774 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 600-14-6 | |
| 100.12 | |
| TZMFJUDUGYTVRY-UHFFFAOYSA-N | |
| 11747 | |
| pentane-2,3-dione |
| C5H8O2 | |
| MFCD00009313 | |
| 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone | |
| CHEBI:52774 | |
| CCC(=O)C(C)=O |
Safety and Handling
EINECSNumber : (2)-0563&(2)-2474
RTECSNumber : SA1850000
TSCA : Yes
Recommended Storage : Freezer