accessibility menu, dialog, popup

UserName

CH3OCH3O

CAS RN 600-14-6

IUPAC Name: pentane-2,3-dione
Synonyms: acetyl propionyl acetylpropionyl
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: TZMFJUDUGYTVRY-UHFFFAOYSA-N
SMILES: CCC(=O)C(C)=O
Percent Purity 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)