missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pentanedione 97.0+%, TCI America™

Catalog No. p-7134395
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
P005125ML 25 mL
P0051100ML 100 mL
P0051500ML 500 mL
3 options

Catalog No. P005125ML

Supplier: TCI America P005125ML

Only null left

Chemical Identifiers

CAS 600-14-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009313
InChI Key TZMFJUDUGYTVRY-UHFFFAOYSA-N
Synonym 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone
PubChem CID 11747
ChEBI CHEBI:52774
IUPAC Name pentane-2,3-dione
SMILES CCC(=O)C(C)=O

Specifications

Melting Point -52°C
Color Yellow
Boiling Point 108°C
Quantity 25 mL
UN Number 1224
Formula Weight 100.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Pentanedione
EINECSNumber (2)-0563&(2)-2474
RTECSNumber SA1850000
TSCA Yes
Recommended Storage Freezer