missing translation for 'onlineSavingsMsg'
Learn More

2,3-O-Isopropylidene-D-glyceraldehyde, 50% w/w in dichloromethane

CAS: 15186-48-8 | C6H10O3 | 130.143 g/mol

Supplier:  Thermo Scientific Chemicals J6633709

 View more versions of this product

Catalog No. AAJ6633709


Only null left
Explore available promotions

Description

Description

Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2,3-O-Isopropylidene-D-glyceraldehyde
Colorless to Yellow
C6H10O3
MFCD00269682
1593
Difficult to mix.
+53.8° (c=2 in Chloroform)
(4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
259712
50% w/w in dichloromethane
15186-48-8
62°C (144°F)
1.454
10 g
r-+-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 4r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-+-glyceraldehyde acetonide, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, r, acetone d-glyceraldehyde, +-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, 4r, 4r-2,2-dimethyl-1,3-dioxolan-4-carbaldehyd, pubchem5767
YSGPYVWACGYQDJ-YFKPBYRVSA-N
CC1(OCC(O1)C=O)C
130.143
130.14
Liquid

Chemical Identifiers

15186-48-8
130.143
YSGPYVWACGYQDJ-YFKPBYRVSA-N
259712
CC1(OCC(O1)C=O)C
C6H10O3
MFCD00269682
r-+-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 4r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-+-glyceraldehyde acetonide, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, r, acetone d-glyceraldehyde, +-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, 4r, 4r-2,2-dimethyl-1,3-dioxolan-4-carbaldehyd, pubchem5767
(4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H227
Suspected of causing cancer.
Combustible liquid.

P201-P202-P210-P235-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H336-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only