accessibility menu, dialog, popup

UserName

CAS RN 15186-48-8

IUPAC Name: (4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
Synonyms: (4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: YSGPYVWACGYQDJ-YFKPBYRVSA-N
SMILES: CC1(C)OC[C@@H](O1)C=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)