missing translation for 'onlineSavingsMsg'
Learn More

2,3-O-Isopropylidene-D-glyceraldehyde, 50% w/w in dichloromethane

Catalog No. p-7043223
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6633703 1 g
AAJ6633706 5 g
AAJ6633709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6633703 Supplier Thermo Scientific Chemicals Supplier No. J6633703
Only null left

CAS: 15186-48-8 | C6H10O3 | 130.143 g/mol

Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 15186-48-8
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00269682
InChI Key YSGPYVWACGYQDJ-YFKPBYRVSA-N
Synonym r-+-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 4r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-+-glyceraldehyde acetonide, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, r, acetone d-glyceraldehyde, +-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, 4r, 4r-2,2-dimethyl-1,3-dioxolan-4-carbaldehyd, pubchem5767
PubChem CID 259712
IUPAC Name (4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
SMILES CC1(OCC(O1)C=O)C

Specifications

Color Colorless to Yellow
Flash Point 62°C (144°F)
Refractive Index 1.454
Quantity 1 g
UN Number UN1593
Solubility Information Difficult to mix.
Optical Rotation +53.8° (c=2 in Chloroform)
Formula Weight 130.14
Concentration or Composition (by Analyte or Components) 50% w/w in dichloromethane
Physical Form Liquid
Chemical Name or Material 2,3-O-Isopropylidene-D-glyceraldehyde
Show More Show Less
Hazard Category H227-H315-H319-H336-H350
Hazard Statement GHS H Statement
H351-H227
Suspected of causing cancer.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only