missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Naphthalenedialdehyde 99.0+%, TCI America™
Supplier: TCI America A55941G
Specifications
| 2,3-Naphthalenedialdehyde [Fluorimetric Reagent for Primary Amines] | |
| 133°C | |
| C12H8O2 | |
| 1 g | |
| ZIPLKLQPLOWLTM-UHFFFAOYSA-N | |
| naphthalene-2,3-dicarbaldehyde | |
| 96400 | |
| ≥99.0% (GC) |
| 7149-49-7 | |
| Yellow | |
| MFCD00016618 | |
| 2,3-naphthalenedicarboxaldehyde, 2,3-naphthalenedialdehyde, naphthalene-2,3-dicarboxaldehyde, 2,3-naphthalenedicarbaldehyde, naphthalene-2,3-dialdehyde, 2,3-nda, 2,3-diformylnaphthalene, 2,3-naphthalene dicarboxaldehyde, nnaphthalene-2,3-dicarboxaldehyde, naphthalene-2,3-dicarboxaldehyde nda | |
| C1=CC=C2C=C(C(=CC2=C1)C=O)C=O | |
| 184.194 | |
| 184.19 | |
| Crystalline Powder |
Chemical Identifiers
| 7149-49-7 | |
| 184.194 | |
| ZIPLKLQPLOWLTM-UHFFFAOYSA-N | |
| 96400 | |
| C1=CC=C2C=C(C(=CC2=C1)C=O)C=O |
| C12H8O2 | |
| MFCD00016618 | |
| 2,3-naphthalenedicarboxaldehyde, 2,3-naphthalenedialdehyde, naphthalene-2,3-dicarboxaldehyde, 2,3-naphthalenedicarbaldehyde, naphthalene-2,3-dialdehyde, 2,3-nda, 2,3-diformylnaphthalene, 2,3-naphthalene dicarboxaldehyde, nnaphthalene-2,3-dicarboxaldehyde, naphthalene-2,3-dicarboxaldehyde nda | |
| naphthalene-2,3-dicarbaldehyde |
Safety and Handling
TSCA : No