accessibility menu, dialog, popup

UserName

OO

CAS RN 7149-49-7

IUPAC Name: naphthalene-2,3-dicarbaldehyde
Synonyms: naphthalene-2,3-dicarbaldehyde
Molecular Weight (g/mol): 184.19
Molecular Formula: C12H8O2
InChi Key: ZIPLKLQPLOWLTM-UHFFFAOYSA-N
SMILES: O=CC1=C(C=O)C=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)