missing translation for 'onlineSavingsMsg'
Learn More

2,3-Naphthalenedialdehyde 99.0+%, TCI America™

Catalog No. p-7118289
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5594100MG 100 mg
A55941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5594100MG Supplier TCI America Supplier No. A5594100MG
Only null left

Chemical Identifiers

CAS 7149-49-7
Molecular Formula C12H8O2
Molecular Weight (g/mol) 184.194
MDL Number MFCD00016618
InChI Key ZIPLKLQPLOWLTM-UHFFFAOYSA-N
Synonym 2,3-naphthalenedicarboxaldehyde, 2,3-naphthalenedialdehyde, naphthalene-2,3-dicarboxaldehyde, 2,3-naphthalenedicarbaldehyde, naphthalene-2,3-dialdehyde, 2,3-nda, 2,3-diformylnaphthalene, 2,3-naphthalene dicarboxaldehyde, nnaphthalene-2,3-dicarboxaldehyde, naphthalene-2,3-dicarboxaldehyde nda
PubChem CID 96400
IUPAC Name naphthalene-2,3-dicarbaldehyde
SMILES C1=CC=C2C=C(C(=CC2=C1)C=O)C=O

Specifications

Melting Point 133°C
Color Yellow
Quantity 100 mg
Formula Weight 184.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Naphthalenedialdehyde [Fluorimetric Reagent for Primary Amines]