missing translation for 'onlineSavingsMsg'
Learn More

2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]benzimidazole 98.0+%, TCI America™

Catalog No. M28001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28001G 1 g
M28005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28001G Supplier TCI America Supplier No. M28001G

Chemical Identifiers

CAS 103577-40-8
Molecular Formula C16H14F3N3OS
Molecular Weight (g/mol) 353.363
MDL Number MFCD00834357
InChI Key CCHLMSUZHFPSFC-UHFFFAOYSA-N
Synonym lansoprazole sulfide, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridin-2-yl methyl thio-1h-benzo d imidazole, 2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methylthio-1h-benzimidazole, unii-58xp950e2t, lansoprazole sulphide compound, 2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methyl thio-1h-benzimidazole, 2-methyl-4-2,2,2-trifluoroethoxy pyridinyl thio-1h-binzimidazole, 1h-benzimidazole,2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methyl thio, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridin-2-yl methyl sulfanyl-1h-1,3-benzodiazole, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridinyl methyl thio-1h-benzimidazole
PubChem CID 1094080
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfanyl]-1H-benzimidazole
SMILES CC1=C(C=CN=C1CSC2=NC3=CC=CC=C3N2)OCC(F)(F)F

Specifications

Melting Point 149°C
Quantity 1 g
Formula Weight 353.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]benzimidazole
TSCA No
Recommended Storage Freezer