accessibility menu, dialog, popup

UserName

CAS RN 103577-40-8

IUPAC Name: 2-({[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methyl}sulfanyl)-1H-1,3-benzodiazole
Synonyms: 2-({[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methyl}sulfanyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 353.36
Molecular Formula: C16H14F3N3OS
InChi Key: CCHLMSUZHFPSFC-UHFFFAOYSA-N
SMILES: CC1=C(OCC(F)(F)F)C=CN=C1CSC1=NC2=CC=CC=C2N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)