missing translation for 'onlineSavingsMsg'
Learn More

2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole, 98%

Catalog No. p-7041371
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61878MD 250 mg
AAH6187803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61878MD Supplier Thermo Scientific Chemicals Supplier No. H61878MD
Only null left

CAS: 103577-40-8 | C16H14F3N3OS | 353.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103577-40-8
Molecular Formula C16H14F3N3OS
Molecular Weight (g/mol) 353.363
MDL Number MFCD00834357
InChI Key CCHLMSUZHFPSFC-UHFFFAOYSA-N
Synonym lansoprazole sulfide, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridin-2-yl methyl thio-1h-benzo d imidazole, 2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methylthio-1h-benzimidazole, unii-58xp950e2t, lansoprazole sulphide compound, 2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methyl thio-1h-benzimidazole, 2-methyl-4-2,2,2-trifluoroethoxy pyridinyl thio-1h-binzimidazole, 1h-benzimidazole,2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methyl thio, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridin-2-yl methyl sulfanyl-1h-1,3-benzodiazole, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridinyl methyl thio-1h-benzimidazole
PubChem CID 1094080
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfanyl]-1H-benzimidazole
SMILES CC1=C(C=CN=C1CSC2=NC3=CC=CC=C3N2)OCC(F)(F)F

Specifications

Color White
Quantity 250 mg
Formula Weight 353.36
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only