missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methoxyphenethyl)phenol 98.0+%, TCI America™

Catalog No. M24721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24721G 1 g
M24725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24721G Supplier TCI America Supplier No. M24721G
Only null left

Chemical Identifiers

CAS 167145-13-3
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.291
MDL Number MFCD09800498
InChI Key HGQQRAXOBYWKDV-UHFFFAOYSA-N
Synonym 2-[2-(3-Methoxyphenyl)ethyl]phenol
PubChem CID 10585448
IUPAC Name 2-[2-(3-methoxyphenyl)ethyl]phenol
SMILES COC1=CC=CC(=C1)CCC2=CC=CC=C2O

Specifications

Melting Point 47°C
Color White
Quantity 1 g
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Methoxyphenethyl)phenol