accessibility menu, dialog, popup

UserName

CH3OOH

CAS RN 167145-13-3

IUPAC Name: 2-[2-(3-methoxyphenyl)ethyl]phenol
Synonyms: 2-[2-(3-methoxyphenyl)ethyl]phenol
Molecular Weight (g/mol): 228.29
Molecular Formula: C15H16O2
InChi Key: HGQQRAXOBYWKDV-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CCC2=CC=CC=C2O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results