missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Fluorophenyl)ethanol 98.0+%, TCI America™

Catalog No. F07761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07761G 1 g
F07765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07761G Supplier TCI America Supplier No. F07761G

Chemical Identifiers

CAS 52059-53-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00045998
InChI Key MZNBGEKFZCWVES-UHFFFAOYSA-N
Synonym 2-3-fluorophenyl ethanol, 3-fluorophenethyl alcohol, 2-3-fluorophenyl ethan-1-ol, benzeneethanol, 3-fluoro, 3-fluorophenethylalcohol, 2-3-fluorophenyl ethylalcohol, 3-fluorobenzenethanol, 3-fluorophenylethanol, 3-fluorobenzeneethanol, acmc-209kxl
PubChem CID 573132
IUPAC Name 2-(3-fluorophenyl)ethan-1-ol
SMILES OCCC1=CC=CC(F)=C1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 1 g
Formula Weight 140.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Fluorophenyl)ethanol