accessibility menu, dialog, popup

UserName

OHF

CAS RN 52059-53-7

IUPAC Name: 2-(3-fluorophenyl)ethan-1-ol
Synonyms: 2-(3-fluorophenyl)ethanol
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: MZNBGEKFZCWVES-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC(F)=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)