missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzaldehyde 96.0+%, TCI America™

Catalog No. p-7127742
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D34631G 1 g
D34635G 5 g
2 options

Catalog No. D34631G

Supplier: TCI America D34631G

Only null left

Chemical Identifiers

CAS 5779-93-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00798004
InChI Key UIFVCPMLQXKEEU-UHFFFAOYSA-N
Synonym benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde
PubChem CID 34224
IUPAC Name 2,3-dimethylbenzaldehyde
SMILES CC1=CC=CC(=C1C)C=O

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
Formula Weight 134.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylbenzaldehyde